Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl-5-amino-3-methyl-2,4-thiophenedicarboxylate 98%

Supplier: AMBEED, INC
Warning

A653885-100G A653885-25G A653885-5G
76980-834EA 155 USD InStock
76980-834 76980-836 76980-838
Diethyl-5-amino-3-methyl-2,4-thiophenedicarboxylate 98%
Diethyl-5-amino-3-methyl-2,4-thiophenedicarboxylate
Precaución: for Research Use Only


Formula: C₁₁H₁₅NO₄S
MW: 257.30 g/mol
MDL Number: MFCD00005450
CAS Number: 4815-30-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty