Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Methoxy-9-oxo-9,10-dihydroacridine-4-carboxylic acid 95%

Supplier: AMBEED, INC
Warning

A110274-100MG A110274-1G A110274-250MG
76970-706EA 19 USD InStock
76970-706 76970-708 76970-710
5-Methoxy-9-oxo-9,10-dihydroacridine-4-carboxylic acid 95%
5-Methoxy-9-oxo-9,10-dihydroacridine-4-carboxylic acid
Precaución: for Research Use Only


Formula: C₁₅H₁₁NO₄
MDL Number: MFCD09955193
CAS Number: 88377-31-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty