Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Nitro-4-quinolinol 97%

Supplier: AMBEED, INC
Warning

A260739-100G A260739-1G A260739-25G A260739-500G A260739-5G A260739-10G
76976-948EA 235 USD InStock
76976-948 76976-950 76976-952 76976-954 76976-956 77672-330
3-Nitro-4-quinolinol 97%
3-Nitro-4-quinolinol
Precaución: for Research Use Only


Formula: C₉H₆N₂O₃
MDL Number: MFCD00277722
CAS Number: 50332-66-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty