Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 3-bromo-4,5-dichlorobenzoate 95%

Supplier: AMBEED, INC
Warning

A236475-100MG A236475-250MG
76976-346EA 9 USD InStock
76976-346 76976-348
Ethyl 3-bromo-4,5-dichlorobenzoate 95%
Ethyl 3-bromo-4,5-dichlorobenzoate
Precaución: for Research Use Only


Formula: C₉H₇BrCl₂O₂
MDL Number: MFCD11846096
CAS Number: 1160574-85-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty