Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-m-phenylenediamine 97%

Supplier: AMBEED, INC
Warning

A152859-1G A152859-5G A152859-25G
76973-168EA 10 USD InStock
76973-168 76973-170 77671-138
4-Chloro-m-phenylenediamine 97%
4-Chloro-m-phenylenediamine
Precaución: for Research Use Only


Formula: C₆H₇ClN₂
MDL Number: MFCD00025284
CAS Number: 5131-60-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty