Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-4-hydroxyphenylboronic acid 98%

Supplier: AMBEED, INC
Warning

A171638-100MG A171638-1G A171638-250MG A171638-5G
76974-024EA 24 USD InStock
76974-024 76974-026 76974-028 77671-442
2-Chloro-4-hydroxyphenylboronic acid 98%
2-Chloro-4-hydroxyphenylboronic acid
Precaución: for Research Use Only


Formula: C₆H₆BClO₃
MDL Number: MFCD02684320
CAS Number: 766549-26-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty