Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Dichlorobenzoic acid 95%

Supplier: AMBEED, INC
Warning

A103197-100G A103197-10G A103197-25G A103197-500G
76970-258EA 36 USD InStock
76970-258 76970-260 76970-262 76970-264
3,4-Dichlorobenzoic acid 95%
3,4-Dichlorobenzoic acid
Precaución: for Research Use Only


Formula: C₇H₄Cl₂O₂
MDL Number: MFCD00002492
CAS Number: 51-44-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty