Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-3-(2-bromophenyl)propionate 97%

Supplier: AMBEED, INC
Warning

A264061-100G A264061-1G
76977-030EA 197 USD InStock
76977-030 76977-032
Methyl-3-(2-bromophenyl)propionate 97%
Methyl-3-(2-bromophenyl)propionate
Precaución: for Research Use Only


Formula: C₁₀H₁₁BrO₂
MDL Number: MFCD09953149
CAS Number: 66191-86-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty