Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Fluoro-m-toluic acid 97%

Supplier: AMBEED, INC
Warning

A153693-10G A153693-1G A153693-250MG A153693-25G A153693-5G
76973-218EA 27 USD InStock
76973-218 76973-220 76973-222 76973-224 76973-226
5-Fluoro-m-toluic acid 97%
5-Fluoro-m-toluic acid
Precaución: for Research Use Only


Formula: C₈H₇FO₂
MDL Number: MFCD03094313
CAS Number: 518070-19-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty