Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Hydroxy-4-methoxy-2-nitrobenzoic acid 97%

Supplier: AMBEED, INC
Warning

A440214-10G A440214-1G A440214-250MG
76979-368EA 10 USD InStock
76979-368 76979-370 76979-372
5-Hydroxy-4-methoxy-2-nitrobenzoic acid 97%
5-Hydroxy-4-methoxy-2-nitrobenzoic acid
Precaución: for Research Use Only


Formula: C₈H₇NO₆
MW: 213.15 g/mol
MDL Number: MFCD10666941
CAS Number: 31839-20-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty