Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pipemidic acid 98%

Supplier: AMBEED, INC
Warning

A311707-100G A311707-25G A311707-500G A311707-5G
76977-916EA 245 USD InStock
76977-916 76977-918 76977-920 76977-922
Pipemidic acid 98%
Pipemidic acid
Precaución: for Research Use Only


Formula: C₁₄H₁₇N₅O₃
MW: 303.322 g/mol
MDL Number: MFCD00057291
CAS Number: 51940-44-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty