Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-3-methoxypyridine hydrochloride 97%

Supplier: AMBEED, INC
Warning

A283179-100MG A283179-1G A283179-250MG
76977-470EA 20 USD InStock
76977-470 76977-472 77672-464
4-Bromo-3-methoxypyridine hydrochloride 97%
4-Bromo-3-methoxypyridine hydrochloride
Precaución: for Research Use Only


Formula: C₆H₇BrClNO
MDL Number: MFCD11226828
CAS Number: 1209335-53-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty