Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-5-bromopicolinamide 96%

Supplier: AMBEED, INC
Warning

A127150-100MG A127150-1G A127150-250MG A127150-5G
76971-916EA 6 USD InStock
76971-916 76971-918 76971-920 77670-796
3-Amino-5-bromopicolinamide 96%
3-Amino-5-bromopicolinamide
Precaución: for Research Use Only


Formula: C₆H₆BrN₃O
MDL Number: MFCD11110235
CAS Number: 669066-89-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty