Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methoxy-trans-β-styrylboronic acid pinacol ester 98%

Supplier: AMBEED, INC
Warning

A346478-1G A346478-250MG A346478-5G
76978-472EA 67 USD InStock
76978-472 76978-474 76978-476
4-Methoxy-trans-β-styrylboronic acid pinacol ester 98%
4-Methoxy-trans-beta-styrylboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₅H₂₁BO₃
MDL Number: MFCD04038966
CAS Number: 149777-83-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty