Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dibromosuccinic acid 98%

Supplier: AMBEED, INC
Danger

A385804-25G A385804-100G
76978-868EA 20 USD InStock
76978-868 77672-910
2,3-Dibromosuccinic acid 98%
2,3-Dibromosuccinic acid
Precaución: for Research Use Only


Formula: HOOC(BrCH)₂COOH
MDL Number: MFCD00004209
CAS Number: 526-78-3
UN: 3261
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty