Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Thenoic acid 98%

Supplier: AMBEED, INC
Warning

A103725-1000G A103725-100G A103725-10G A103725-25G A103725-500G
76970-292EA 49 USD InStock
76970-292 76970-294 76970-296 76970-298 76970-300
2-Thenoic acid 98%
2-Thenoic acid
Precaución: for Research Use Only


Formula: C₅H₄O₂S
MW: 128.15 g/mol
Boiling Pt: 260
MDL Number: MFCD00005437
CAS Number: 527-72-0
Merck Index: 12,09491
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty