Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

PLX4032 98+%

Supplier: AMBEED, INC
Warning

A116840-1G A116840-10MG A116840-100MG A116840-250MG A116840-50MG
76971-260EA 320 USD InStock
76971-260 77610-958 77670-640 77670-642 77670-644
PLX4032 98+%
PLX4032
Precaución: for Research Use Only


Formula: C₂₃H₁₈ClF₂N₃O₃S
MDL Number: MFCD18074504
CAS Number: 918504-65-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty