Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Methyl-1,3-dihydrofuro[3,4-c]pyridin-7-ol 95%

Supplier: AMBEED, INC
Danger

A170526-100MG A170526-250MG
76973-986EA 119 USD InStock
76973-986 76973-988
6-Methyl-1,3-dihydrofuro[3,4-c]pyridin-7-ol 95%
6-Methyl-1,3-dihydrofuro[3,4-c]pyridin-7-ol
Precaución: For research use only.


Formula: C₈H₉NO₂
MDL Number: MFCD00460998
CAS Number: 5196-20-3
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty