Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

cis-2-(tert-Butyl)maleonitrile 98%

Supplier: AMBEED, INC
Danger

A774407-10G A774407-1G A774407-25G A774407-5G
76981-538EA 63 USD InStock
76981-538 76981-540 76981-542 76981-544
cis-2-(tert-Butyl)maleonitrile 98%
cis-2-(tert-Butyl)maleonitrile
Precaución: for Research Use Only


Formula: C₈H₁₀N₂
MDL Number: MFCD12405597
CAS Number: 169309-80-2
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty