Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-Fluorophenyl)thiophene 95%

Supplier: AMBEED, INC
Warning

A137830-100G A137830-10G A137830-1G A137830-25G A137830-500G A137830-5G
76972-414EA 59 USD InStock
76972-414 76972-416 76972-418 76972-420 76972-422 76972-424
2-(4-Fluorophenyl)thiophene 95%
2-(4-Fluorophenyl)thiophene
Precaución: for Research Use Only


Formula: C₁₀H₇FS
MDL Number: MFCD06802535
CAS Number: 58861-48-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty