Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzenemethanol 97%

Supplier: AMBEED, INC
Warning

A357377-100MG A357377-10G A357377-1G A357377-250MG A357377-5G
76978-556EA 6 USD InStock
76978-556 76978-558 76978-560 76978-562 76978-564
2-Methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzenemethanol 97%
2-Methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzenemethanol
Precaución: for Research Use Only


Formula: C₁₄H₂₁BO₄
MW: 264.12 g/mol
MDL Number: MFCD13184694
CAS Number: 1009303-77-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty