Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(2-Naphthyl)benzeneboronic acid 98%

Supplier: AMBEED, INC
Warning

A247675-10G A247675-1G A247675-250MG A247675-25G A247675-5G
76976-608EA 203 USD InStock
76976-608 76976-610 76976-612 76976-614 76976-616
3-(2-Naphthyl)benzeneboronic acid 98%
3-(2-Naphthyl)benzeneboronic acid
Precaución: for Research Use Only


Formula: C₁₆H₁₃BO₂
MDL Number: MFCD16293738
CAS Number: 870774-29-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty