Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzothiazol-4-ylamine 95%

Supplier: AMBEED, INC
Warning

A415321-100MG A415321-1G A415321-250MG A415321-5G
76979-202EA 39 USD InStock
76979-202 76979-204 76979-206 76979-208
Benzothiazol-4-ylamine 95%
Benzothiazol-4-ylamine
Precaución: for Research Use Only


Formula: C₇H₆N₂S
MDL Number: MFCD00672135
CAS Number: 1123-51-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty