Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 5-bromo-1-methyl-1H-pyrazole-4-carboxylate 95%

Supplier: AMBEED, INC
Warning

A155081-100MG A155081-1G A155081-250MG A155081-5G
76973-280EA 10 USD InStock
76973-280 76973-282 76973-284 77671-192
Ethyl 5-bromo-1-methyl-1H-pyrazole-4-carboxylate 95%
Ethyl 5-bromo-1-methyl-1H-pyrazole-4-carboxylate
Precaución: for Research Use Only


Formula: C₇H₉BrN₂O₂
MDL Number: MFCD13189831
CAS Number: 105486-72-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty