Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,6-Dichloro-2-(methylthio)-5-nitropyrimidine 98%

Supplier: AMBEED, INC
Warning

A231111-10G A231111-1G A231111-250MG A231111-25G A231111-5G
76976-158EA 84 USD InStock
76976-158 76976-160 76976-162 76976-164 76976-166
4,6-Dichloro-2-(methylthio)-5-nitropyrimidine 98%
4,6-Dichloro-2-(methylthio)-5-nitropyrimidine
Precaución: for Research Use Only


Formula: C₅H₃Cl₂N₃O₂S
MDL Number: MFCD02323208
CAS Number: 1979-96-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty