Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2'-((4-Aminophenyl)azanediyl)diethanol sulfate 97%

Supplier: AMBEED, INC
Warning

A791184-1G A791184-25G A791184-5G A791184-500G
76981-616EA 8 USD InStock
76981-616 76981-618 76981-620 77674-150
2,2'-((4-Aminophenyl)azanediyl)diethanol sulfate 97%
2,2'-((4-Aminophenyl)azanediyl)diethanol sulfate
Precaución: for Research Use Only


Formula: C₁₀H₁₈N₂O₆S
MW: 294.33 g/mol
MDL Number: MFCD00013177
CAS Number: 54381-16-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty