Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Hydroxybenzylacetone 98%

Supplier: AMBEED, INC
Warning

A108894-100G A108894-500G
76970-572EA 17 USD InStock
76970-572 76970-574 76970-576
4-Hydroxybenzylacetone 98%
4-Hydroxybenzylacetone
Precaución: for Research Use Only


Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 292
MDL Number: MFCD00002394
CAS Number: 5471-51-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty