Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(Pyridin-3-yl)piperazine hydrochloride 95%

Supplier: AMBEED, INC
Warning

A196433-100MG A196433-1G A196433-250MG
76975-064EA 32 USD InStock
76975-064 76975-066 76975-068
1-(Pyridin-3-yl)piperazine hydrochloride 95%
1-(Pyridin-3-yl)piperazine hydrochloride
Precaución: for Research Use Only


Formula: C₉H₁₄ClN₃
MDL Number: MFCD21396210
CAS Number: 1010133-97-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty