Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dichloro-2-iodopyrazine 98%

Supplier: AMBEED, INC
Warning

A619358-100MG A619358-10G A619358-1G A619358-250MG A619358-5G
76980-604EA 17 USD InStock
76980-604 76980-606 76980-608 76980-610 76980-612
3,5-Dichloro-2-iodopyrazine 98%
3,5-Dichloro-2-iodopyrazine
Precaución: for Research Use Only


Formula: C4HCl2IN2
MDL Number: MFCD13189860
CAS Number: 136866-30-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty