Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cbz-L-arginine hydrochloride 98%

Supplier: AMBEED, INC
Warning

A514551-100G A514551-25G A514551-5G
76979-950EA 92 USD InStock
76979-950 76979-952 76979-954
Cbz-L-arginine hydrochloride 98%
Cbz-L-arginine hydrochloride
Precaución: for Research Use Only


Formula: C₁₄H₂₁ClN₄O₄
MW: 344.796 g/mol
MDL Number: MFCD00038946
CAS Number: 56672-63-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty