Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(2-Methoxyethyl)phenol 97%

Supplier: AMBEED, INC
Warning

A521252-100G A521252-25G A521252-500G A521252-5G
76980-012EA 20 USD InStock
76980-012 76980-014 76980-016 76980-018
4-(2-Methoxyethyl)phenol 97%
4-(2-Methoxyethyl)phenol
Precaución: for Research Use Only


Formula: C₉H₁₂O₂
MW: 152.19 g/mol
MDL Number: MFCD00017537
CAS Number: 56718-71-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty