Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H-Benzo[d]imidazole-4-carbonitrile 95%

Supplier: AMBEED, INC
Warning

A153116-100MG A153116-250MG
76973-194EA 23 USD InStock
76973-194 76973-196
1H-Benzo[d]imidazole-4-carbonitrile 95%
1H-Benzo[d]imidazole-4-carbonitrile
Precaución: for Research Use Only


Formula: C₈H₅N₃
MDL Number: MFCD18204825
CAS Number: 64574-21-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty