Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Imidazo[1,2-a]pyridine-8-carbonitrile 95%

Supplier: AMBEED, INC
Danger

A536558-1G A536558-250MG
76980-130EA 107 USD InStock
76980-130 76980-132
Imidazo[1,2-a]pyridine-8-carbonitrile 95%
Imidazo[1,2-a]pyridine-8-carbonitrile
Precaución: for Research Use Only


Formula: C₈H₅N₃
MDL Number: MFCD09994637
CAS Number: 136117-70-9
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty