Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Chloro-5-fluoropicolinic acid 97%

Supplier: AMBEED, INC
Warning

A169889-10G A169889-1G A169889-250MG A169889-25G A169889-5G
76973-946EA 161 USD InStock
76973-946 76973-948 76973-950 76973-952 76973-954
6-Chloro-5-fluoropicolinic acid 97%
6-Chloro-5-fluoropicolinic acid
Precaución: for Research Use Only


Formula: C₆H₃ClFNO₂
MDL Number: MFCD13185819
CAS Number: 860296-24-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty