Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bis-trispropane 95%

Supplier: AMBEED, INC
Warning

A100332-100G A100332-10G A100332-25G A100332-5G
77025-114EA 67 USD InStock
77025-114 77025-116 77025-118 77025-120
Bis-trispropane 95%
Bis-trispropane
Precaución: for Research Use Only


Formula: C₁₁H₂₆N₂O₆
MW: 282.34 g/mol
MDL Number: MFCD00004689
CAS Number: 64431-96-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty