Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-4-(chloromethyl)pyridine hydrochloride 98%

Supplier: AMBEED, INC
Danger

A100339-100MG A100339-250MG A100339-5G
77025-122EA 22 USD InStock
77025-122 77025-124 77025-126
2-Amino-4-(chloromethyl)pyridine hydrochloride 98%
2-Amino-4-(chloromethyl)pyridine hydrochloride
Precaución: for Research Use Only


Formula: C₆H₇ClN₂·ClH
MDL Number: MFCD12198864
CAS Number: 1186663-31-9
UN: 3261
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty