Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Pyrrolidin-1-ylsulfonyl)benzeneboronic acid 97%

Supplier: AMBEED, INC
Warning

A103286-100MG A103286-1G A103286-250MG
76994-646EA 25 USD InStock
76994-646 76994-648 76994-650
2-(Pyrrolidin-1-ylsulfonyl)benzeneboronic acid 97%
2-(Pyrrolidin-1-ylsulfonyl)benzeneboronic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₄BNO₄S
MW: 255.098 g/mol
MDL Number: MFCD08689515
CAS Number: 913835-83-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty