Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-4-(2-methoxyethoxy)benzene 97%

Supplier: AMBEED, INC
Warning

A103646-10G A103646-1G A103646-250MG A103646-5G
76982-308EA 310 USD InStock
76982-308 76982-310 76982-312 76982-314
1-Bromo-4-(2-methoxyethoxy)benzene 97%
1-Bromo-4-(2-methoxyethoxy)benzene
Precaución: for Research Use Only


Formula: C₉H₁₁BrO₂
MW: 231.1 g/mol
MDL Number: MFCD09931307
CAS Number: 39255-23-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty