Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methyl-1H-indazole 98%

Supplier: AMBEED, INC
Warning

A103713-10G A103713-1G A103713-250MG A103713-25G A103713-5G
76982-316EA 92 USD InStock
76982-316 76982-318 76982-320 76982-322 76982-324
3-Methyl-1H-indazole 98%
3-Methyl-1H-indazole
Precaución: for Research Use Only


Formula: C₈H₈N₂
MDL Number: MFCD00601361
CAS Number: 3176-62-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty