Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,6-Dichloro-3-methylpyridazine 90%

Supplier: AMBEED, INC
Warning

A103793-100MG A103793-250MG
76982-328EA 69 USD InStock
76982-328 76982-330
4,6-Dichloro-3-methylpyridazine 90%
4,6-Dichloro-3-methylpyridazine
Precaución: for Research Use Only


Formula: C₅H₄Cl₂N₂
MDL Number: MFCD21604745
CAS Number: 68240-43-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty