Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Rebastinib (DCC-2036) 98%

Supplier: AMBEED, INC
Warning

A104117-10MG A104117-1MG A104117-50MG A104117-5MG
77008-518EA 54 USD InStock
77008-518 77008-520 77008-522 77008-524
Rebastinib (DCC-2036) 98%
Rebastinib (DCC-2036)
Precaución: for Research Use Only


Formula: C₃₀H₂₈FN₇O₃
MDL Number: MFCD19443646
CAS Number: 1020172-07-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty