Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(3,6-Dimethoxy-2-nitrophenyl)ethanone 95%

Supplier: AMBEED, INC
Warning

A104411-100MG A104411-1G A104411-250MG
77008-528EA 19 USD InStock
77008-528 77008-530 77008-532
1-(3,6-Dimethoxy-2-nitrophenyl)ethanone 95%
1-(3,6-Dimethoxy-2-nitrophenyl)ethanone
Precaución: for Research Use Only


Formula: C₁₀H₁₁NO₅
MW: 225.20 g/mol
MDL Number: MFCD18431633
CAS Number: 90923-05-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty