Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Carbobenzoxy-D-phenylalanine 97%

Supplier: AMBEED, INC
Warning

A104852-10G A104852-1G A104852-5G A104852-25G
77025-372EA 11 USD InStock
77025-372 77025-374 77025-376 77570-286
N-Carbobenzoxy-D-phenylalanine 97%
N-Carbobenzoxy-D-phenylalanine
Precaución: for Research Use Only


Formula: C₁₇H₁₇NO₄
MW: 299.33 g/mol
MDL Number: MFCD00063151
CAS Number: 2448-45-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty