Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

GKT-137831 97%

Supplier: AMBEED, INC
Warning

A104916-100MG A104916-10MG A104916-1MG A104916-5MG
76982-368EA 67 USD InStock
76982-368 76982-370 76982-372 76982-374
GKT-137831 97%
GKT-137831
Precaución: for Research Use Only


Formula: C₂₁H₂₀ClN₄O₂
MDL Number: MFCD27923122
CAS Number: 1218942-37-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty