Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methylcinnamic acid 98% predominantly trans

Supplier: AMBEED, INC
Warning

A105172-100G A105172-1G A105172-25G A105172-5G
76994-824EA 187 USD InStock
76994-824 76994-826 76994-828 76994-830
2-Methylcinnamic acid 98% predominantly trans
2-Methylcinnamic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₀O₂
MDL Number: MFCD00016841
CAS Number: 2373-76-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty