Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-4-fluorophenylacetonitrile 98%

Supplier: AMBEED, INC
Warning

A105433-10G A105433-100G
76994-864EA 23 USD InStock
76994-864 77670-516
2-Bromo-4-fluorophenylacetonitrile 98%
2-Bromo-4-fluorophenylacetonitrile
Precaución: for Research Use Only


Formula: C₈H₅BrFN
MW: 214.04 g/mol
MDL Number: MFCD04115829
CAS Number: 61150-58-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty