Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-5-methoxybenzamide 95%

Supplier: AMBEED, INC
Warning

A105524-1G A105524-250MG
77008-582EA 38.2 USD InStock
77008-582 77008-584 77008-586
3-Amino-5-methoxybenzamide 95%
3-Amino-5-methoxybenzamide
Precaución: for Research Use Only


Formula: C₈H₁₀N₂O₂
MDL Number: MFCD18389709
CAS Number: 960234-99-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty