Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Crizotinib 98%

Supplier: AMBEED, INC
Danger

A106537-10MG A106537-1G A106537-1MG A106537-250MG A106537-50MG A106537-5G A106537-25G
77025-466EA 6 USD InStock
77025-466 77025-468 77025-470 77025-472 77025-474 77025-476 77666-554
Crizotinib 98%
Crizotinib
Precaución: for Research Use Only


Formula: C₂₁H₂₂Cl₂FN₅O
MDL Number: MFCD12407409
CAS Number: 877399-52-5
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty