Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-2-thenoic acid 98%

Supplier: AMBEED, INC
Warning

A107858-100G A107858-10G A107858-25G A107858-500G A107858-5G
76982-500EA 24 USD InStock
76982-500 76982-502 76982-504 76982-506 76982-508
5-Chloro-2-thenoic acid 98%
5-Chloro-2-thenoic acid
Precaución: for Research Use Only


Formula: C₅H₃ClO₂S
MW: 162.6 g/mol
MDL Number: MFCD00041426
CAS Number: 24065-33-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty