Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,6-Diiodo-9-phenylcarbazole 97%

Supplier: AMBEED, INC
Warning

A107914-10G A107914-1G A107914-250MG A107914-5G
76982-528EA 57 USD InStock
76982-528 76982-530 76982-532 76982-534
3,6-Diiodo-9-phenylcarbazole 97%
3,6-Diiodo-9-phenylcarbazole
Precaución: for Research Use Only


Formula: C₁₈H₁₁I₂N
MDL Number: MFCD11521285
CAS Number: 57103-21-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty